Learn More
Accuflow Systems Saint-Gobain FEP Tubing, 5/32 ID x 1/4 OD, 50 ft
Catalog No. NC2327989
Encompass_Preferred
FEP tubing with 5/32 in inner diameter and 1/4 in outer diameter, 0.047 in wall thickness, supplied in a 50 ft coil. The tubing offers broad chemical compatibility and a wide operating temperature range suitable for laboratory, pharmaceutical, bioprocessing, and semiconductor fluid-transfer applications.
  • FEP fluoropolymer construction provides chemical inertness and low extractables.
  • 5/32 in inner diameter, 1/4 in outer diameter, 0.047 in wall thickness.
  • Supplied as a 50 ft length coil for continuous runs or dispensing.
  • Wide temperature range: -328°F to 402°F (-200°C to 206°C).
  • Resistant to most solvents, acids, and fuels.
  • UV resistant for extended outdoor or light-exposed use.
Catalog No. NC2327989 Supplier Accuflow Systems Supplier No. TSFE14025004750
FEP tubing with 5/32 in inner diameter and 1/4 in outer diameter, 0.047 in wall thickness, supplied in a 50 ft coil. The tubing offers broad chemical compatibility and a wide operating temperature range suitable for laboratory, pharmaceutical, bioprocessing, and semiconductor fluid-transfer applications.
  • FEP fluoropolymer construction provides chemical inertness and low extractables.
  • 5/32 in inner diameter, 1/4 in outer diameter, 0.047 in wall thickness.
  • Supplied as a 50 ft length coil for continuous runs or dispensing.
  • Wide temperature range: -328°F to 402°F (-200°C to 206°C).
  • Resistant to most solvents, acids, and fuels.
  • UV resistant for extended outdoor or light-exposed use.