Learn More
SAI Infusion Technologies Micro500 500uL Blood Tube- Lithium Heparin, Light Green Push Cap
SDP
Catalog No. NC0985898
Encompass
500ul Micro500 Blood Tube - Lithium Heparin, Light Green Cap, may be used for Plasma collection. SAI's Micro500 blood tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only.
Catalog No. NC0985898 Supplier SAI Infusion Technologies Supplier No. M500H
500ul Micro500 Blood Tube - Lithium Heparin, Light Green Cap, may be used for Plasma collection. SAI's Micro500 blood tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only.