Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, A-60-G, 1/4 in ID, 50 ft
Catalog No. 502365853
Encompass_Preferred
Saint-Gobain Tygon A-60-G tubing is an industrial-grade Norprene-formulated tubing engineered for chemical dispensing and fluid-transfer applications. The 1/4 in ID x 1/2 in OD size in a 50 ft length offers high flexural fatigue resistance, wide temperature tolerance, and compatibility with many cleaning and dispensing chemicals.
  • Industrial-grade thermoplastic elastomer construction for chemical resistance.
  • 1/4 in inner diameter, 1/2 in outer diameter, 1/8 in wall, 50 ft length.
  • Temperature range -75°F to 275°F (-60°C to 135°C).
  • Working pressure 19 psi at 73°F, 12 psi at 180°F.
  • Vacuum rating 29.9 inHg at typical temperatures.
  • Suitable for soap, disinfectant, cleaning chemical, and caustic dispensing applications.
Catalog No. 50-236-5853 Supplier Accuflow Systems Supplier No. AFL00019
Saint-Gobain Tygon A-60-G tubing is an industrial-grade Norprene-formulated tubing engineered for chemical dispensing and fluid-transfer applications. The 1/4 in ID x 1/2 in OD size in a 50 ft length offers high flexural fatigue resistance, wide temperature tolerance, and compatibility with many cleaning and dispensing chemicals.
  • Industrial-grade thermoplastic elastomer construction for chemical resistance.
  • 1/4 in inner diameter, 1/2 in outer diameter, 1/8 in wall, 50 ft length.
  • Temperature range -75°F to 275°F (-60°C to 135°C).
  • Working pressure 19 psi at 73°F, 12 psi at 180°F.
  • Vacuum rating 29.9 inHg at typical temperatures.
  • Suitable for soap, disinfectant, cleaning chemical, and caustic dispensing applications.