Learn More
Saint Gobain Performance Plastics TUB NORPRENE 3/8 x 5/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics AFL00029
Encompass_Preferred
Saint-Gobain Norprene Industrial Tubing Formula A-60-G Diameter Inner 3/8 in. Thickness 0.125 in. Overall Length 50 ft. Black
Catalog No. 50-206-9008