Learn More
IDEX Health & Science Upchurch PEEK Tubing 1/32" OD x .020" ID, Orange, 100ft
Catalog No. NC1790596
Encompass_Preferred
Biocompatible, PEEK Tubing is frequently used in high-pressure applications. Its distinct material delivers excellent chemical resistance and is specifically designed to be inert to most solvents. PEEK tubing offers a smooth internal surface that improves the resolution of sample bands.
Catalog No. NC1790596 Supplier IDEX Health & Science Upchurch Supplier No. 1569XL
Biocompatible, PEEK Tubing is frequently used in high-pressure applications. Its distinct material delivers excellent chemical resistance and is specifically designed to be inert to most solvents. PEEK tubing offers a smooth internal surface that improves the resolution of sample bands.