Learn More
Accuflow Systems Saint-Gobain PFA Tubing, 3/16 in I.D., 1/4 in O.D.
Catalog No. NC2327995
Encompass_Preferred
Saint-Gobain Versilon PFA tubing, 3/16 in I.D. x 1/4 in O.D., 0.031 in wall, supplied in 50 ft coils. The tubing offers high chemical resistance, a smooth low-extractables bore, and a wide temperature range suitable for semiconductor, pharmaceutical, and laboratory fluid-transfer applications.
  • Chemically inert fluoropolymer with excellent chemical resistance.
  • Ultra-pure, smooth bore minimizes extractables and particulate generation.
  • Wide operating temperature range: -196°C to 260°C (-320°F to 500°F).
  • Autoclavable and non-stick for easy cleaning and sterilization.
  • 0.031 in wall thickness provides structural stability with flexible handling.
  • Common applications include semiconductor processing, pharmaceutical fluid systems, and analytical instruments.
Catalog No. NC2327995 Supplier Accuflow Systems Supplier No. TSPF35025003150
Saint-Gobain Versilon PFA tubing, 3/16 in I.D. x 1/4 in O.D., 0.031 in wall, supplied in 50 ft coils. The tubing offers high chemical resistance, a smooth low-extractables bore, and a wide temperature range suitable for semiconductor, pharmaceutical, and laboratory fluid-transfer applications.
  • Chemically inert fluoropolymer with excellent chemical resistance.
  • Ultra-pure, smooth bore minimizes extractables and particulate generation.
  • Wide operating temperature range: -196°C to 260°C (-320°F to 500°F).
  • Autoclavable and non-stick for easy cleaning and sterilization.
  • 0.031 in wall thickness provides structural stability with flexible handling.
  • Common applications include semiconductor processing, pharmaceutical fluid systems, and analytical instruments.