Learn More
Accuflow Systems Saint-Gobain Tygon ND 100-65 Tubing, 1/4 in I.D., 100 ft
Catalog No. 502365771
Encompass_Preferred
Tygon ND 100-65 tubing is a medical/surgical-grade flexible tubing formulation designed for reliable fluid transfer in clinical and laboratory equipment. It provides precise, consistent dimensions and flexible handling for applications requiring clarity and predictable performance.
  • Medical/surgical grade formulation for clinical and laboratory fluid handling.
  • 1/4 in inner diameter, 7/16 in outer diameter, 3/32 in wall thickness.
  • 100 ft packaged length for continuous runs and assemblies.
  • Flexible, transparent construction for visual monitoring of fluid flow.
  • Suitable for peristaltic pump applications and general tubing assemblies.
Catalog No. 50-236-5771 Supplier Accuflow Systems Supplier No. ADF02018
Tygon ND 100-65 tubing is a medical/surgical-grade flexible tubing formulation designed for reliable fluid transfer in clinical and laboratory equipment. It provides precise, consistent dimensions and flexible handling for applications requiring clarity and predictable performance.
  • Medical/surgical grade formulation for clinical and laboratory fluid handling.
  • 1/4 in inner diameter, 7/16 in outer diameter, 3/32 in wall thickness.
  • 100 ft packaged length for continuous runs and assemblies.
  • Flexible, transparent construction for visual monitoring of fluid flow.
  • Suitable for peristaltic pump applications and general tubing assemblies.