Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/4 in ID, 7/16 in OD, Non-DEHP
Catalog No. 502365909
Encompass_Preferred
Tygon ND 100-65 medical tubing is a biocompatible, non-DEHP polymer tube engineered for blood-contact and other medical and surgical fluid transfer applications. It provides flexibility, chemical resistance, and consistent processing characteristics for minimally invasive devices, laboratory fluid handling, and medical instrument connections.
  • Biocompatible non-DEHP polymer suitable for blood-contact applications.
  • Inner diameter 1/4 in, outer diameter 7/16 in, wall thickness 3/32 in.
  • Supplied in 50 ft packaged length.
  • Meets USP Class VI and ISO 10993 biocompatibility standards.
  • Flexible and chemically resistant for minimally invasive devices and laboratory use.
  • Compatible with common bonding and welding processes for medical tubing.
Catalog No. 50-236-5909 Supplier Accuflow Systems Supplier No. ADF00018
Tygon ND 100-65 medical tubing is a biocompatible, non-DEHP polymer tube engineered for blood-contact and other medical and surgical fluid transfer applications. It provides flexibility, chemical resistance, and consistent processing characteristics for minimally invasive devices, laboratory fluid handling, and medical instrument connections.
  • Biocompatible non-DEHP polymer suitable for blood-contact applications.
  • Inner diameter 1/4 in, outer diameter 7/16 in, wall thickness 3/32 in.
  • Supplied in 50 ft packaged length.
  • Meets USP Class VI and ISO 10993 biocompatibility standards.
  • Flexible and chemically resistant for minimally invasive devices and laboratory use.
  • Compatible with common bonding and welding processes for medical tubing.