Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Non-DEHP, 3/4 In ID, 1 In OD
Catalog No. 502365769
Encompass_Preferred
Clear, non-DEHP medical/surgical tubing engineered for fluid transfer and medical device applications. The tubing provides consistent dimensional tolerances, biocompatibility, and ease of bonding and welding for reliable performance in peristaltic pumps and surgical fluid handling.
  • Clear for easy visualization of fluid flow.
  • Non-DEHP formulation suitable for medical use.
  • Meets ISO 10993 and relevant USP biocompatibility criteria.
  • Manufactured in a cleanroom environment to medical device standards.
  • 3/4 in inner diameter, 1 in outer diameter, 1/8 in wall thickness.
  • 50 ft package length (supplier packaging).
  • Easy to bond and weld for device assembly and tubing connections.
  • Suitable for peristaltic pump and surgical fluid transfer applications.
Catalog No. 50-236-5769 Supplier Accuflow Systems Supplier No. ADF00053
Clear, non-DEHP medical/surgical tubing engineered for fluid transfer and medical device applications. The tubing provides consistent dimensional tolerances, biocompatibility, and ease of bonding and welding for reliable performance in peristaltic pumps and surgical fluid handling.
  • Clear for easy visualization of fluid flow.
  • Non-DEHP formulation suitable for medical use.
  • Meets ISO 10993 and relevant USP biocompatibility criteria.
  • Manufactured in a cleanroom environment to medical device standards.
  • 3/4 in inner diameter, 1 in outer diameter, 1/8 in wall thickness.
  • 50 ft package length (supplier packaging).
  • Easy to bond and weld for device assembly and tubing connections.
  • Suitable for peristaltic pump and surgical fluid transfer applications.