Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, S-50-HL, 5/8 in ID, 7/8 OD
Catalog No. 502365787
Encompass
Tygon S-50-HL medical tubing is a clear, flexible, medical-grade formulation designed for reliable fluid transfer in blood-contact and peristaltic pump applications. This spool provides 50 ft of 5/8 in inner diameter tubing with a 7/8 in outer diameter, characterized for biocompatibility and compatibility with common sterilization methods.
  • Medical-grade formulation characterized to ISO 10993 series and meets USP Class VI criteria.
  • 5/8 in inner diameter, 7/8 in outer diameter, 1/8 in wall thickness, 50 ft spool.
  • Suitable for blood, IV solutions, dialysis, chemotherapy delivery, and minimally invasive devices.
  • Compatible with EtO, gas, and gamma sterilization methods.
  • Crystal clear for visual monitoring of fluid flow.
  • Non-wetting surface permits complete drainage and reduces residual fluid retention.
  • Demonstrates hemocompatibility and is non-toxic, non-hemolytic, and non-pyrogenic; not intended for implant use.
Catalog No. 50-236-5787 Supplier Accuflow Systems Supplier No. ADF00046
Tygon S-50-HL medical tubing is a clear, flexible, medical-grade formulation designed for reliable fluid transfer in blood-contact and peristaltic pump applications. This spool provides 50 ft of 5/8 in inner diameter tubing with a 7/8 in outer diameter, characterized for biocompatibility and compatibility with common sterilization methods.
  • Medical-grade formulation characterized to ISO 10993 series and meets USP Class VI criteria.
  • 5/8 in inner diameter, 7/8 in outer diameter, 1/8 in wall thickness, 50 ft spool.
  • Suitable for blood, IV solutions, dialysis, chemotherapy delivery, and minimally invasive devices.
  • Compatible with EtO, gas, and gamma sterilization methods.
  • Crystal clear for visual monitoring of fluid flow.
  • Non-wetting surface permits complete drainage and reduces residual fluid retention.
  • Demonstrates hemocompatibility and is non-toxic, non-hemolytic, and non-pyrogenic; not intended for implant use.