Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Medical grade, 1/2 in ID, 50 ft
Catalog No. 502365887
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 medical tubing is a clear, medical-grade thermoplastic tubing designed for reliable fluid transfer in clinical and medical device applications. It provides visual flow monitoring, biocompatibility, and is easy to bond and weld. Supplied in 50 ft lengths, the tubing has a 1/2 in inner diameter, 3/4 in outer diameter, and 1/8 in wall thickness, and is manufactured without the intentional addition of DEHP.
  • Clear tubing for visual flow monitoring.
  • Medical-grade thermoplastic with biocompatibility testing to ISO 10993 and USP standards.
  • No intentional addition of DEHP or animal-derived materials.
  • Dimensions: 1/2 in ID, 3/4 in OD, 1/8 in wall.
  • Supplied in 50 ft packages.
  • Suitable for blood/IV solutions, dialysis, peristaltic pumps, and respiratory therapy.
Catalog No. 50-236-5887 Supplier Accuflow Systems Supplier No. ADF00038
Saint-Gobain Tygon ND 100-65 medical tubing is a clear, medical-grade thermoplastic tubing designed for reliable fluid transfer in clinical and medical device applications. It provides visual flow monitoring, biocompatibility, and is easy to bond and weld. Supplied in 50 ft lengths, the tubing has a 1/2 in inner diameter, 3/4 in outer diameter, and 1/8 in wall thickness, and is manufactured without the intentional addition of DEHP.
  • Clear tubing for visual flow monitoring.
  • Medical-grade thermoplastic with biocompatibility testing to ISO 10993 and USP standards.
  • No intentional addition of DEHP or animal-derived materials.
  • Dimensions: 1/2 in ID, 3/4 in OD, 1/8 in wall.
  • Supplied in 50 ft packages.
  • Suitable for blood/IV solutions, dialysis, peristaltic pumps, and respiratory therapy.