Tygon ND 100-65 is a crystal-clear, medical-grade, non-DEHP flexible tubing formulated with a TOTM plasticizer for use with bodily fluids and medical devices. It is supplied as 50 ft continuous lengths and is suitable for applications requiring USP Class VI biocompatibility and compatibility with common sterilization methods.
- Crystal clear for visual monitoring of fluid flow.
- Non-DEHP formulation (TOTM plasticizer) for reduced plasticizer migration.
- 1/4 in inner diameter, 3/8 in outer diameter, 1/16 in wall thickness.
- Supplied as 50 ft continuous length.
- Maximum suggested working pressure 30 psi at 73°F.
- Minimum bend radius 1 in for routing and handling.
- Vacuum rating 25 in Hg at 73°F.
- Meets USP Class VI and REACH compliance.
- Compatible with EtO, gas, and gamma sterilization methods.
- Non-wetting surface permits complete drainage.