Learn More
Accuflow Systems Saint-Gobain Tygon Medical Tubing, Non-DEHP, 3/16 in I.D., 50 ft
Catalog No. NC3268901
Encompass_Preferred
Tygon ND 100-65 is a clear, biocompatible, non-DEHP medical tubing designed for blood-contact and other medical device applications. It provides visual monitoring of fluid flow, reliable performance in peristaltic pumps, and easy bonding and welding for device assembly.
  • Clear tubing for visualization of fluid flow.
  • Biocompatible, tested to elements of ISO 10993 and USP <88> Class VI.
  • Non-DEHP formulation; manufactured without the intentional addition of DEHP.
  • Dimensions: 3/16 in I.D., 5/16 in O.D., 1/16 in wall.
  • Supplied as a 50 ft roll.
  • Suitable for blood and IV solutions, chemotherapy delivery, dialysis, diagnostic equipment/IVDs, peristaltic pumps, and respiratory therapy.
  • Easy to bond and weld for medical device assembly.
Catalog No. NC3268901 Supplier Accuflow Systems Supplier No. ADF00012
Tygon ND 100-65 is a clear, biocompatible, non-DEHP medical tubing designed for blood-contact and other medical device applications. It provides visual monitoring of fluid flow, reliable performance in peristaltic pumps, and easy bonding and welding for device assembly.
  • Clear tubing for visualization of fluid flow.
  • Biocompatible, tested to elements of ISO 10993 and USP <88> Class VI.
  • Non-DEHP formulation; manufactured without the intentional addition of DEHP.
  • Dimensions: 3/16 in I.D., 5/16 in O.D., 1/16 in wall.
  • Supplied as a 50 ft roll.
  • Suitable for blood and IV solutions, chemotherapy delivery, dialysis, diagnostic equipment/IVDs, peristaltic pumps, and respiratory therapy.
  • Easy to bond and weld for medical device assembly.