Learn More
Saint Gobain Performance Plastics TUB SURG 3/8 x 1/2 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ADF00027
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 Medical Tubings Diameter Inner 3/8 in. Diameter Outer 1/2 in. Thickness 1/16 in. Non-DEHP Overall Length 50 ft. 15.2 m Clear
Catalog No. 50-206-8991