Learn More
Accuflow Systems Saint-Gobain Tygon Medical Tubing, 3/8 in ID, 1/2 in OD
Catalog No. 502365812
Encompass_Preferred
Tygon ND 100-65 medical tubing provides flexible, non-DEHP, medical-grade fluid transfer with a 3/8 in internal diameter, 1/2 in outside diameter, and 1/16 in wall thickness. Supplied in 50 ft lengths, it is biocompatible per ISO 10993 testing and suitable for dialysis, drug delivery, peristaltic pumps, and diagnostic equipment. Maximum recommended operating temperature is 165°F (74°C).
  • 3/8 in internal diameter, 1/2 in outside diameter.
  • 1/16 in wall thickness.
  • Supplied in 50 ft standard length.
  • Non-DEHP medical-grade formulation.
  • Tested to ISO 10993 biocompatibility and USP Class VI.
  • Suitable for peristaltic pumps, dialysis, and drug delivery.
  • Maximum recommended temperature 165°F (74°C).
Catalog No. 50-236-5812 Supplier Accuflow Systems Supplier No. ADF00027
Tygon ND 100-65 medical tubing provides flexible, non-DEHP, medical-grade fluid transfer with a 3/8 in internal diameter, 1/2 in outside diameter, and 1/16 in wall thickness. Supplied in 50 ft lengths, it is biocompatible per ISO 10993 testing and suitable for dialysis, drug delivery, peristaltic pumps, and diagnostic equipment. Maximum recommended operating temperature is 165°F (74°C).
  • 3/8 in internal diameter, 1/2 in outside diameter.
  • 1/16 in wall thickness.
  • Supplied in 50 ft standard length.
  • Non-DEHP medical-grade formulation.
  • Tested to ISO 10993 biocompatibility and USP Class VI.
  • Suitable for peristaltic pumps, dialysis, and drug delivery.
  • Maximum recommended temperature 165°F (74°C).