Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Tygon 2475 Tubing, 1/2 in ID, 50 ft, Plasticizer-Free

Catalog No. 502365852
Encompass

Tygon 2475 is a high-purity, plasticizer-free thermoplastic tubing engineered for low extractables and reduced gas permeation. It meets USP Class VI biocompatibility standards and is suitable for medical, diagnostic, and laboratory fluid transfer applications that require chemical resistance and cleanroom compatibility.

  • Inner diameter: 1/2 in (12.7 mm).
  • Outer diameter: 3/4 in (19.0 mm).
  • Wall thickness: 1/8 in (3.2 mm).
  • Length: 50 ft (15.2 m).
  • Material: Tygon 2475 thermoplastic, plasticizer-free.
  • Color: Clear.
  • Biocompatibility: Meets USP Class VI.
  • Typical applications: diagnostic reagent delivery, medical dispensing, cell and tissue culture transfer, cleanroom fluid lines.

Catalog No. 50-236-5852 Supplier Accuflow Systems Supplier No. ACG00038
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tygon 2475 is a high-purity, plasticizer-free thermoplastic tubing engineered for low extractables and reduced gas permeation. It meets USP Class VI biocompatibility standards and is suitable for medical, diagnostic, and laboratory fluid transfer applications that require chemical resistance and cleanroom compatibility.

  • Inner diameter: 1/2 in (12.7 mm).
  • Outer diameter: 3/4 in (19.0 mm).
  • Wall thickness: 1/8 in (3.2 mm).
  • Length: 50 ft (15.2 m).
  • Material: Tygon 2475 thermoplastic, plasticizer-free.
  • Color: Clear.
  • Biocompatibility: Meets USP Class VI.
  • Typical applications: diagnostic reagent delivery, medical dispensing, cell and tissue culture transfer, cleanroom fluid lines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.