Learn More
Accuflow Systems Saint-Gobain Tygon 2375 Tubing, 1/4 in ID, 3/8 in OD
Catalog No. 502365911
Encompass_Preferred
Tygon 2375 is an ultra chemical-resistant, plasticizer-free thermoplastic tubing engineered for chemical transfer and cleaning applications. It combines excellent chemical resistance, clarity, and flexibility while minimizing fluid contamination and sorption.
  • Ultra chemical resistance to acids, bases, salts, and alcohols.
  • Clear, flexible thermoplastic elastomer, plasticizer-free.
  • Available in 1/4 in ID x 3/8 in OD, 1/16 in wall.
  • Maximum recommended operating temperature 130°F (54°C).
  • Working pressure 25 psi at 73°F for 1/4 in ID x 3/8 in OD.
  • Hardness 75 Shore A.
  • Low sorption and smooth inner surface to minimize contamination.
  • Low-temperature flexibility and impact resistance to -103°F (-75°C).
  • Tested to ASTM methods for pressure, hardness, tensile, and tear.
Catalog No. 50-236-5911 Supplier Accuflow Systems Supplier No. AJK00017
Tygon 2375 is an ultra chemical-resistant, plasticizer-free thermoplastic tubing engineered for chemical transfer and cleaning applications. It combines excellent chemical resistance, clarity, and flexibility while minimizing fluid contamination and sorption.
  • Ultra chemical resistance to acids, bases, salts, and alcohols.
  • Clear, flexible thermoplastic elastomer, plasticizer-free.
  • Available in 1/4 in ID x 3/8 in OD, 1/16 in wall.
  • Maximum recommended operating temperature 130°F (54°C).
  • Working pressure 25 psi at 73°F for 1/4 in ID x 3/8 in OD.
  • Hardness 75 Shore A.
  • Low sorption and smooth inner surface to minimize contamination.
  • Low-temperature flexibility and impact resistance to -103°F (-75°C).
  • Tested to ASTM methods for pressure, hardness, tensile, and tear.