Learn More
Accuflow Systems Saint-Gobain Tygon 2375 Tubing, 5/16 in ID, 7/16 in OD
Catalog No. 502365782
Encompass_Preferred
Tygon 2375 tubing is a plasticizer-free, ultra chemical-resistant flexible tubing designed for demanding chemical transfer and fluid-handling applications. The 5/16 in ID, 7/16 in OD construction with a 1/16 in wall balances flexibility and flow while minimizing sorption and particulate buildup.
  • Plasticizer-free, non-DEHP formulation for chemical resistance.
  • Inner diameter 5/16 in, outer diameter 7/16 in, wall thickness 1/16 in.
  • Clear tubing with smooth inner surface to reduce particulate buildup.
  • Working pressure 20 psi at 73°F.
  • Minimum bend radius 1.25 in.
  • Maximum recommended operating temperature 130 °F (54 °C); flexible to -103 °F (-75 °C).
  • Excellent resistance to acids, bases, salts, and alcohols; fair resistance to ketones.
  • Low sorption to minimize cross-contamination.
  • Vacuum rating 29.9 inHg.
  • Durometer Shore A 75; tensile strength 1,900 psi; ultimate elongation 850%.
Catalog No. 50-236-5782 Supplier Accuflow Systems Supplier No. AJK00022
Tygon 2375 tubing is a plasticizer-free, ultra chemical-resistant flexible tubing designed for demanding chemical transfer and fluid-handling applications. The 5/16 in ID, 7/16 in OD construction with a 1/16 in wall balances flexibility and flow while minimizing sorption and particulate buildup.
  • Plasticizer-free, non-DEHP formulation for chemical resistance.
  • Inner diameter 5/16 in, outer diameter 7/16 in, wall thickness 1/16 in.
  • Clear tubing with smooth inner surface to reduce particulate buildup.
  • Working pressure 20 psi at 73°F.
  • Minimum bend radius 1.25 in.
  • Maximum recommended operating temperature 130 °F (54 °C); flexible to -103 °F (-75 °C).
  • Excellent resistance to acids, bases, salts, and alcohols; fair resistance to ketones.
  • Low sorption to minimize cross-contamination.
  • Vacuum rating 29.9 inHg.
  • Durometer Shore A 75; tensile strength 1,900 psi; ultimate elongation 850%.