Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, High Purity, 1/4 in I.D., 50 ft
Catalog No. 502365838
Encompass_Preferred
Tygon 2475 high-purity tubing is a crystal-clear, plasticizer-free thermoplastic tubing designed for medical, diagnostic, and biopharmaceutical fluid transfer applications. It provides low gas permeation and a smooth inner bore to minimize particle retention and reduce variability in sensitive fluids.
  • Plasticizer-free construction reduces extractables and leachables.
  • Smooth inner surface limits protein and particle retention.
  • Low gas permeability reduces moisture loss and oxidation.
  • Clear, flexible thermoplastic suitable for sterile fluid transfer.
  • Meets USP Class VI biocompatibility standards.
  • Available in 1/4 in I.D. x 3/8 in O.D., 1/16 in wall, 50 ft length.
Catalog No. 50-236-5838 Supplier Accuflow Systems Supplier No. ACG00017
Tygon 2475 high-purity tubing is a crystal-clear, plasticizer-free thermoplastic tubing designed for medical, diagnostic, and biopharmaceutical fluid transfer applications. It provides low gas permeation and a smooth inner bore to minimize particle retention and reduce variability in sensitive fluids.
  • Plasticizer-free construction reduces extractables and leachables.
  • Smooth inner surface limits protein and particle retention.
  • Low gas permeability reduces moisture loss and oxidation.
  • Clear, flexible thermoplastic suitable for sterile fluid transfer.
  • Meets USP Class VI biocompatibility standards.
  • Available in 1/4 in I.D. x 3/8 in O.D., 1/16 in wall, 50 ft length.