Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Plasticizer-Free, 1/8 in ID, 50 ft
Catalog No. 502365908
Encompass_Preferred
Tygon 2475 tubing is a crystal-clear, plasticizer-free thermoplastic designed for high-purity medical, diagnostic, and biopharmaceutical fluid transfer. It provides low gas permeation, reduced leachables, and a smooth inner bore to minimize particle and protein retention, making it well suited for reagent delivery, dispensing, and cleanroom transfer applications.
  • Plasticizer-free thermoplastic reduces leachables and contamination risk.
  • Low gas permeation protects moisture-sensitive fluids from oxidation and moisture loss.
  • Meets USP Class VI biocompatibility for medical and diagnostic use.
  • Smooth inner bore limits protein and particle retention.
  • Transparent material allows visual monitoring of flow and air bubbles.
  • 50 ft length, 1/8 in inner diameter, 1/4 in outer diameter, 1/16 in wall thickness.
Catalog No. 50-236-5908 Supplier Accuflow Systems Supplier No. ACG00007
Tygon 2475 tubing is a crystal-clear, plasticizer-free thermoplastic designed for high-purity medical, diagnostic, and biopharmaceutical fluid transfer. It provides low gas permeation, reduced leachables, and a smooth inner bore to minimize particle and protein retention, making it well suited for reagent delivery, dispensing, and cleanroom transfer applications.
  • Plasticizer-free thermoplastic reduces leachables and contamination risk.
  • Low gas permeation protects moisture-sensitive fluids from oxidation and moisture loss.
  • Meets USP Class VI biocompatibility for medical and diagnostic use.
  • Smooth inner bore limits protein and particle retention.
  • Transparent material allows visual monitoring of flow and air bubbles.
  • 50 ft length, 1/8 in inner diameter, 1/4 in outer diameter, 1/16 in wall thickness.