Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 3/8 in ID, 50 ft
Catalog No. NC2390178
Encompass_Preferred
Saint-Gobain Tygon 2475 high-purity thermoplastic tubing (3/8 in ID x 1/2 in OD, 1/16 in wall, 50 ft) provides low extractables, USP Class VI biocompatibility, and reduced gas permeation for reliable medical and diagnostic fluid transfer.
  • Plasticizer-free thermoplastic formulation for low extractables and chemical resistance.
  • 3/8 in inner diameter, 1/2 in outer diameter, 1/16 in wall thickness.
  • 50 ft packaged length for system routing and replacement stock.
  • Smooth inner bore reduces protein and particle retention.
  • Meets USP Class VI biocompatibility requirements.
  • Working pressure approximately 20 psi at 73°F for this size.
  • Crystal-clear transparency for visual flow monitoring.
Catalog No. NC2390178 Supplier Accuflow Systems Supplier No. ACG00027
Saint-Gobain Tygon 2475 high-purity thermoplastic tubing (3/8 in ID x 1/2 in OD, 1/16 in wall, 50 ft) provides low extractables, USP Class VI biocompatibility, and reduced gas permeation for reliable medical and diagnostic fluid transfer.
  • Plasticizer-free thermoplastic formulation for low extractables and chemical resistance.
  • 3/8 in inner diameter, 1/2 in outer diameter, 1/16 in wall thickness.
  • 50 ft packaged length for system routing and replacement stock.
  • Smooth inner bore reduces protein and particle retention.
  • Meets USP Class VI biocompatibility requirements.
  • Working pressure approximately 20 psi at 73°F for this size.
  • Crystal-clear transparency for visual flow monitoring.