Learn More
Accuflow Systems Saint-Gobain Tygon 2475 Tubing, High-Purity, 5/16 in ID, 50 ft
Catalog No. 502365851
Encompass_Preferred
Saint-Gobain Tygon 2475 high-purity tubing is a plasticizer-free thermoplastic designed for sensitive fluid transfer applications. It provides high clarity, minimal absorption and adsorption, and a smooth inner surface to minimize particulate entrapment. The material meets USP Class VI biological test requirements and is compatible with radiation and ethylene oxide sterilization for use in sterile filling, diagnostics, and laboratory instrumentation.
  • High-purity, plasticizer-free thermoplastic formulation.
  • Inner diameter 5/16 in, outer diameter 7/16 in, wall thickness 1/16 in, 50 ft length.
  • High clarity for visual monitoring of fluid flow.
  • Low gas permeation and exceptionally low absorption/adsorption.
  • Smooth inner surface reduces particulate entrapment and contamination risk.
  • Sterilizable by radiation and ethylene oxide.
Catalog No. 50-236-5851 Supplier Accuflow Systems Supplier No. ACG00022
Saint-Gobain Tygon 2475 high-purity tubing is a plasticizer-free thermoplastic designed for sensitive fluid transfer applications. It provides high clarity, minimal absorption and adsorption, and a smooth inner surface to minimize particulate entrapment. The material meets USP Class VI biological test requirements and is compatible with radiation and ethylene oxide sterilization for use in sterile filling, diagnostics, and laboratory instrumentation.
  • High-purity, plasticizer-free thermoplastic formulation.
  • Inner diameter 5/16 in, outer diameter 7/16 in, wall thickness 1/16 in, 50 ft length.
  • High clarity for visual monitoring of fluid flow.
  • Low gas permeation and exceptionally low absorption/adsorption.
  • Smooth inner surface reduces particulate entrapment and contamination risk.
  • Sterilizable by radiation and ethylene oxide.