Learn More
Saint Gobain Performance Plastics TUB TYGN 1-1/4 x 1-1/2 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACF00069
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 1-1/4 in. Diameter Outer 1-1/2 in. Maximum Pressure 13 psi at 73 deg.F Thickness 1/8 in. Clear
Catalog No. 50-206-8972