Learn More
Enzo Life Sciences alpha-Tubulin Recombinant monoclonal antibody (F2C) (Rabbit IgG lambda) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6100200
Encompass_Preferred
Clone: F2C. Isotype: IgGλ. Immunogen: Human MBP (microtubule-binding protein). Species reactivity: Human. Applications: ELISA, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3136