Learn More
Aviva Systems Biology Corp TUBA1C Antibody - C-terminal region (ARP60613_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60613_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBA1C. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TUBA1C. Alternate names: MGC10851, MGC14580, TUBA6, bcm948ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-103