Learn More
Aviva Systems Biology Corp TUBA1C Antibody - C-terminal region (ARP60614_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60614_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBA1C. It was validated on Western Blot. . Alternate names: MGC10851, MGC14580, TUBA6, bcm948ApplicationIHC, IP, WB. Species reactivity: Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-287-104