Learn More
Aviva Systems Biology Corp TUBA3C Antibody - N-terminal region (ARP53650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBA3C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TUBA3C. Alternate names: TUBA2, bA408E5.3, TUBA3DApplicationWB. Species reactivity: Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53724