Learn More
Cayman Chemical Tubacin, 537049-40-4, 1 mg
SDP
Supplier:  Cayman Chemical 136911
Encompass_Preferred
Molecular Formula C41H43N3O7S, Purity ≥90%, Formula Weight 721.9; A selective HDAC6 inhibitor (IC50 = 4 nM) used as a biochemical tool to control microtubule-dependent intracellular trafficking, to manipulate the aggresome formation of misfolded proteins in certain diseases, and to study the dynamics of cellular adhesion.
Catalog No. NC0778559