Learn More
Medchemexpress LLC HY-13428 1mg Medchemexpress, Tubacin CAS:537049-40-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134281MG
Encompass_Preferred
Medchemexpress, HY-13428 1mg Tubacin CAS:537049-40-4 Tubacin is a potent and selective inhibitor of HDAC6, with an IC50 value of 4 nM and approximately 350-fold selectivity over HDAC1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8993