Learn More
Enzo Life Sciences Tubacin (500µg). CAS: 537049-40-4
SDP
Supplier:  Enzo Life Sciences BMLGR3620500
Encompass_Preferred
Tubacin (tubulin acetylation inducer) is a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6 (IC50=0.004µM). IC50's for the other HDACs are 1000-fold higher, making tubacin both more selective and more potent than Tubastatin A, which also inhibits HDAC8. Concentration in cell culture experiments typically ranges from 2-50µM. Alternative name: N-(4-{(2R,4R,6S)-4-{[(4,5-diphenyl-1,3-oxazol-2-yl)sulfanyl]methyl}-6-[4-(hydroxymethyl)phenyl]-1,3-dioxan-2-yl}phenyl)-N'-hydroxyoctanediamide. Purity: ≥96% (ELSD). Solubility: Soluble in DMSO (10 mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-1013