Learn More
Selleck Chemical LLC Tubastatin A 50mg 1252003-15-8
SDP
Catalog No. NC0769182
Encompass_Preferred
Tubastatin A is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay. It is selective against all the other isozymes (1000-fold) except HDAC8 (57-fold). Tubastatin A promotes autophagy and increases apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0769182 Supplier Selleck Chemical LLC Supplier No. S804950MG
Tubastatin A is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay. It is selective against all the other isozymes (1000-fold) except HDAC8 (57-fold). Tubastatin A promotes autophagy and increases apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.