Learn More
Medchemexpress LLC HY-13271A 200mg , Tubastatin-A CAS:1252003-15-8 Purity:98%
SDP
Catalog No. 501492627
Encompass_Preferred
Medchemexpress, HY-13271A 200mg Tubastatin-A CAS:1252003-15-8 Formula:C20H21N3O2 IC50: 15 nM (HDAC6) Purity:98% Tubastatin A is a potent and selective  HDAC6  inhibitor with  IC 50  of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2627 Supplier Medchemexpress LLC Supplier No. HY13271A200MG
Medchemexpress, HY-13271A 200mg Tubastatin-A CAS:1252003-15-8 Formula:C20H21N3O2 IC50: 15 nM (HDAC6) Purity:98% Tubastatin A is a potent and selective  HDAC6  inhibitor with  IC 50  of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.