Learn More
Apexbio Technology LLC Tubastatin A, 50mg. Cas: 1252003-15-8 MFCD: MFCD18071463. HDAC6 inhibitor,potent and selective
SDP
Catalog No. 501904880
Encompass_Preferred
Tubastatin A is a potent and selective inhibitor of HDAC6 with IC50 value of 15 nM [1]. Histone deacetylases (HDACs) and histone acetyltransferases (HATs) mediates the balance between histone deacetylation and acetylation. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4880 Supplier Apexbio Technology LLC Supplier No. A410150
Tubastatin A is a potent and selective inhibitor of HDAC6 with IC50 value of 15 nM [1]. Histone deacetylases (HDACs) and histone acetyltransferases (HATs) mediates the balance between histone deacetylation and acetylation. Other sizes are available. Please inqury us for quote.