Learn More
Selleck Chemical LLC Tubastatin A HCl-10mg
SDP
Supplier:  Selleck Chemical LLC S262710MG
Encompass_Preferred
Tubastatin A HCl (TSA) is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay. It is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more).
Catalog No. 50-238-9972