Learn More
Active Motif Tubastatin A hydrochloride
SDP
Catalog No. NC1840903
Encompass
Active Motif's Potent and selective histone deacetylase 6 (HDAC 6) inhibitor (IC50 = 15 nm; >1000-fold selective against other HDACs and 60-fold for HDAC8)). Inhibits TNF_ (IC50 = 272 nm) and IL-6 (IC50 = 712 nm) in LPS-stimulated human macrophages and displays anti-inflammatory activity in animal models. Improves survival in animal models of sepsis. Improves cognition in Alzheimers disease transgenic mice. It is cell permeable.
Catalog No. NC1840903 Supplier Active Motif Supplier No. 14085
Active Motif's Potent and selective histone deacetylase 6 (HDAC 6) inhibitor (IC50 = 15 nm; >1000-fold selective against other HDACs and 60-fold for HDAC8)). Inhibits TNF_ (IC50 = 272 nm) and IL-6 (IC50 = 712 nm) in LPS-stimulated human macrophages and displays anti-inflammatory activity in animal models. Improves survival in animal models of sepsis. Improves cognition in Alzheimers disease transgenic mice. It is cell permeable.