Learn More
TARGETMOL CHEMICALS INC TUBASTATIN A HYDROCHLORI 100MG
SDP
Catalog No. 502576587
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubastatin A Hydrochloride (TSA HCl) is an effective and specific HDAC6 inhibitor (IC50 15 nM). It has selectivity (>1000-fold) against all other isozymes except HDAC8 (>57-fold). purity: 99%
Catalog No. 50-257-6587 Supplier TARGETMOL CHEMICALS INC Supplier No. T6161100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubastatin A Hydrochloride (TSA HCl) is an effective and specific HDAC6 inhibitor (IC50 15 nM). It has selectivity (>1000-fold) against all other isozymes except HDAC8 (>57-fold). purity: 99%