Learn More
Cayman Chemical Tubastatin A trifluoroacetat
SDP
Catalog No. 502890226
Encompass_Preferred
Potent HDAC6 inhibitor (IC50 = 15 nM) with 1,000-fold selectivity against all other HDAC isoforms (IC50s >16 μM), excluding HDAC8 (IC50 = 0.9 μM); induces α-tubulin hyperacetylation at 2.5 μM in primary cortical neuron cultures; displays dose-dependent neuronal protection of primary cortical neuron cultures at 5-10 μM
Catalog No. 50-289-0226 Supplier Cayman Chemical Supplier No. 1055910 MG
Potent HDAC6 inhibitor (IC50 = 15 nM) with 1,000-fold selectivity against all other HDAC isoforms (IC50s >16 μM), excluding HDAC8 (IC50 = 0.9 μM); induces α-tubulin hyperacetylation at 2.5 μM in primary cortical neuron cultures; displays dose-dependent neuronal protection of primary cortical neuron cultures at 5-10 μM