Learn More
Aviva Systems Biology Corp TUBB Antibody - N-terminal region (ARP55761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TUBB. Alternate names: M40, MGC117247, MGC16435, OK/SW-cl.56, TUBB1, TUBB5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61602