Learn More
Aviva Systems Biology Corp TUBB2A Antibody - middle region (ARP40177_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40177_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBB2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TUBB2A. Alternate names: TUBB, TUBB2, dJ40E16.7ApplicationIHC, WB. Species reactivity: Human, Mouse, Rat, Sheep
Catalog No. 50-014-57321