Learn More
Aviva Systems Biology Corp TUBB3 Antibody - C-terminal region (ARP63783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBB3. It was validated on Western Blot. . Alternate names: CFEOM3A, MC1R, TUBB4, beta-4, CDCBMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-288-083