Learn More
Aviva Systems Biology Corp TUBB6 Antibody - N-terminal region (ARP60382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBB6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TUBB6. Alternate names: HsT1601, MGC132410, MGC4083, TUBB-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-027