Learn More
Andwin Scientific TUBBY CONTAINER 5/PK
SDP
Catalog No. NC0660164
Encompass_Preferred
This 330x200x115mm polypropylene t7ubby stackable storage container is ideal for keeping tubes, tips, pipets, gloves and other small items organized. With the removable dividers, big or small compartments can be created as needed. It features a lid that snaps in place to secure contents. Tubby containers are proportionally sized to utilize space efficiently when stacked, with same footprint but twice as deep.
Catalog No. NC0660164 Supplier Andwin Scientific Supplier No. HD23453
This 330x200x115mm polypropylene t7ubby stackable storage container is ideal for keeping tubes, tips, pipets, gloves and other small items organized. With the removable dividers, big or small compartments can be created as needed. It features a lid that snaps in place to secure contents. Tubby containers are proportionally sized to utilize space efficiently when stacked, with same footprint but twice as deep.