Learn More
Thomas Scientific RT PCR Tube, 0.1mL, 8-strip, Flat Cap, White, Sterile, 125/pk
SDP
Catalog No. 501457383
Encompass_Preferred
RT PCR Tube, 0.1ml, 8-strip, Flat Cap, White, sterile 125/pk Rapid, even, and accurate heat transfer for reliable Real-time amplifications No-leak tight seal Certified DNase, RNase, DNA, PCR inhibitor, and pyrogen free
Catalog No. 50-145-7383 Supplier Thomas Scientific Supplier No. 1159V06
RT PCR Tube, 0.1ml, 8-strip, Flat Cap, White, sterile 125/pk Rapid, even, and accurate heat transfer for reliable Real-time amplifications No-leak tight seal Certified DNase, RNase, DNA, PCR inhibitor, and pyrogen free