Learn More
Thomas Scientific RT PCR Tube, 0.2mL 12-strip, Flat Cap, White, Sterile, 80/pk
SDP
Catalog No. 501457384
Encompass_Preferred
RT PCR Tube, 0.2mL 12-strip, Flat Cap, White, Sterile 80/pk Rapid, even, and accurate heat transfer for reliable Real-time amplifications No-leak tight seal Certified DNase, RNase, DNA, PCR inhibitor, and pyrogen free
Catalog No. 50-145-7384 Supplier Thomas Scientific Supplier No. 1159V05
RT PCR Tube, 0.2mL 12-strip, Flat Cap, White, Sterile 80/pk Rapid, even, and accurate heat transfer for reliable Real-time amplifications No-leak tight seal Certified DNase, RNase, DNA, PCR inhibitor, and pyrogen free