Learn More
USA Scientific Inc 1.5 mL copolymer microcentrifuge tube, amber
Catalog No. NC9700554
Encompass_Preferred
1.5 mL copolymer polypropylene graduated microcentrifuge tube, amber, 500/bag
Catalog No. NC9700554 Supplier USA Scientific Inc Supplier No. 14152507
1.5 mL copolymer polypropylene graduated microcentrifuge tube, amber, 500/bag