Learn More
CELLTREAT Scientific Products TUBE ONLY, 1.5mL Screw Top Micro Tube, Conical Bottom, Graduated, Non-sterile, 1000/CS
SDP
Catalog No. 501535135
Encompass_Preferred
CELLTREAT Scientific Products 1.5mL Screw Top Micro Tube Only, Conical Bottom, Graduated. High temperature resistivity range--shatter/crack resistant and ideal for boiling. Easy 3/4 turn to open and close. Caps are available separately in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Grip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation. Manufactured from high clarity virgin polypropylene. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Max RCF: 20,000g. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 1,000/case.
Catalog No. 50-153-5135 Supplier CELLTREAT Scientific Products Supplier No. 230823
CELLTREAT Scientific Products 1.5mL Screw Top Micro Tube Only, Conical Bottom, Graduated. High temperature resistivity range--shatter/crack resistant and ideal for boiling. Easy 3/4 turn to open and close. Caps are available separately in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Grip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation. Manufactured from high clarity virgin polypropylene. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Max RCF: 20,000g. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 1,000/case.