Learn More
Ace Glass, Inc. Tube, reaction/storage, 100mL, 24/40 standard taper neck, 0-3 inlet valve side arm, No-Air
SDP
Catalog No. 50975357
Encompass_Preferred
100mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 24/40 standard taper neck, and a 0-3 inlet valve side arm.
Catalog No. 50-975-357 Supplier Ace Glass, Inc. Supplier No. 7752215
100mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 24/40 standard taper neck, and a 0-3 inlet valve side arm.