Learn More
Thomas Scientific 10mL Centr. Tube w/screw cap, sterile
SDP
Catalog No. 501458821
Encompass
10mL centrifuge tube, polypropylene (PP) with screw cap and printed graduations Dimensions: 16 x 100mm Available in non-sterile and sterile packaging White graduation marks and writing area Non-sterile tube (#6292) has natural screw cap Sterile tube (#6293) has yellow screw cap Spin Rate: 3000 RCF
This item is not returnable. View return policy
Catalog No. 50-145-8821 Supplier Thomas Scientific Supplier No. 1216M93
10mL centrifuge tube, polypropylene (PP) with screw cap and printed graduations Dimensions: 16 x 100mm Available in non-sterile and sterile packaging White graduation marks and writing area Non-sterile tube (#6292) has natural screw cap Sterile tube (#6293) has yellow screw cap Spin Rate: 3000 RCF