Learn More
Labcon North America SuperClear 17x100mm Culture Tubes with Dual Position Caps, Polystyrene, 25 per Bag, Sterile
Catalog No. 50101559
Encompass_Preferred
Tube, Culture; Labcon; SuperClear; 14mL; Sterile; 17x100mm culture tubes with attached dual position polyethylene caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 14mL; 25 tubes per bag/20 bags per case; 17mm W. 100mm H; Made in USA
Catalog No. 50-101-559 Supplier Labcon North America Supplier No. 35353350009
Tube, Culture; Labcon; SuperClear; 14mL; Sterile; 17x100mm culture tubes with attached dual position polyethylene caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 14mL; 25 tubes per bag/20 bags per case; 17mm W. 100mm H; Made in USA