Learn More
Ace Glass, Inc. Tube, pressure, 120mL, 150psi, 38.1mm OD, 17.8cm long, complete with #25 back-seal plug
SDP
Catalog No. 50976195
Encompass_Preferred
120mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Includes #25 back-seal plug. Dimensions: 38.1mm O.D. x 17.8cm long.
Catalog No. 50-976-195 Supplier Ace Glass, Inc. Supplier No. 8648110
120mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Includes #25 back-seal plug. Dimensions: 38.1mm O.D. x 17.8cm long.